Assay Details
| Gene Symbol: 1700015G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195601.1 | NP_001182530.1 | 1 - 2 | 75 | 92 | |
| GenBank mRNA | AK005994.1 | - | 1 - 2 | 78 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small VCP/p97-interacting protein | Svip |
1700006C13Rik, 6620401M08Rik |
-8.8kb | Sense | Chr. 7 - 51997161 - 52006018 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 33898 |
Gm33898 | 871.9kb | Anti-sense | Chr. 7 - 52887595 - 52976998 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001195601.1 transcripts.
Set Membership: |
|

1700015G11Rik
Genomic Map