Assay Details
| Gene Symbol: Ano5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271879.1 | NP_001258808.1 | 3 - 4 | 326 | 86 | |
| NR_073508.1 | 4 - 5 | 361 | 86 | |||
| GenBank mRNA | AK034197.1 | - | 4 - 5 | 350 | 86 | |
| BC109163.2 | - | 3 - 4 | 248 | 40047987 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ano5
Genomic Map