Assay Details
| Gene Symbol: Ano5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271879.1 | NP_001258808.1 | 4 - 5 | 426 | 100 | |
| NM_177694.6 | NP_808362.2 | 5 - 6 | 468 | 100 | ||
| NR_073508.1 | 5 - 6 | 461 | 100 | |||
| GenBank mRNA | AB125740.1 | - | 5 - 6 | 468 | 100 | |
| AB206762.1 | - | 5 - 6 | 468 | 100 | ||
| AK034197.1 | - | 5 - 6 | 450 | 100 | ||
| BC109163.2 | - | 4 - 5 | 348 | 40047987 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ano5
Genomic Map