Assay Details
| Gene Symbol: Ranbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019930.2 | NP_064314.2 | 10 - 11 | 1656 | 148 | |
| GenBank mRNA | AF006465.1 | - | 11 - 12 | 1485 | 148 | |
| AK132714.1 | - | 10 - 11 | 1135 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ranbp9
Genomic Map