Assay Details
| Gene Symbol: Ranbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019930.2 | NP_064314.2 | 12 - 13 | 1932 | 94 | |
| GenBank mRNA | AF006465.1 | - | 13 - 14 | 1761 | 94 | |
| AK132714.1 | - | 12 - 13 | 1411 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ranbp9
Genomic Map