Assay Details
| Gene Symbol: Ippk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276399.1 | NP_001263328.1 | 10 - 11 | 1304 | 102 | |
| NM_199056.3 | NP_951011.1 | 10 - 11 | 1223 | 102 | ||
| NR_075085.1 | 11 - 12 | 1308 | 102 | |||
| GenBank mRNA | AK029085.1 | - | 10 - 11 | 1170 | 102 | |
| AK031987.1 | - | 10 - 11 | 1304 | 102 | ||
| AK160092.1 | - | 10 - 11 | 1170 | 102 | ||
| AK167467.1 | - | 10 - 11 | 1198 | 102 | ||
| BC024630.1 | - | 11 - 12 | 1253 | 4222376 | 102 | |
| BC062167.1 | - | 10 - 11 | 1135 | 6517021 | 102 | |
| BC065157.1 | - | 10 - 11 | 1138 | 6416865 | 102 | |
| DV647168.1 | - | 2 - 3 | 338 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ippk
Genomic Map