Assay Details
| Gene Symbol: Ippk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276399.1 | NP_001263328.1 | 4 - 5 | 536 | 91 | |
| NM_199056.3 | NP_951011.1 | 4 - 5 | 455 | 91 | ||
| NR_075085.1 | 5 - 6 | 540 | 91 | |||
| GenBank mRNA | AK031987.1 | - | 4 - 5 | 537 | 91 | |
| AK160092.1 | - | 4 - 5 | 402 | 91 | ||
| AK167467.1 | - | 4 - 5 | 430 | 91 | ||
| BC024630.1 | - | 5 - 6 | 485 | 4222376 | 91 | |
| BC062167.1 | - | 4 - 5 | 367 | 6517021 | 91 | |
| BC065157.1 | - | 4 - 5 | 370 | 6416865 | 91 | |
| CD353694.1 | - | 1 - 2 | 81 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ippk
Genomic Map