Assay Details
| Gene Symbol: Vim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011701.4 | NP_035831.2 | 5 - 6 | 1129 | 71 | |
| GenBank mRNA | AK012469.1 | - | 5 - 6 | 632 | 71 | |
| AK076001.1 | - | 5 - 6 | 1126 | 71 | ||
| AK131618.1 | - | 5 - 6 | 1131 | 71 | ||
| AK133726.1 | - | 5 - 6 | 1130 | 71 | ||
| AK150270.1 | - | 5 - 6 | 825 | 71 | ||
| AK153019.1 | - | 5 - 6 | 1132 | 71 | ||
| AK159539.1 | - | 5 - 6 | 1130 | 71 | ||
| AK169181.1 | - | 5 - 6 | 1131 | 71 | ||
| AK170429.1 | - | 5 - 6 | 1130 | 71 | ||
| AK213831.1 | - | 1 - 2 | 57 | 71 | ||
| BC089335.1 | - | 5 - 6 | 1125 | 6415629 | 71 | |
| M24849.1 | - | 5 - 6 | 1046 | 71 | ||
| M26251.1 | - | 5 - 6 | 1488 | 71 | ||
| X56397.1 | - | 5 - 6 | 1092 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vim
Genomic Map