Assay Details
| Gene Symbol: Tmem87b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028248.2 | NP_082524.2 | 12 - 13 | 1247 | 132 | |
| GenBank mRNA | AK050203.1 | - | 12 - 13 | 1239 | 132 | |
| AK171811.1 | - | 13 - 14 | 1163 | 132 | ||
| AK172611.1 | - | 13 - 14 | 1163 | 132 | ||
| BC031488.1 | - | 12 - 13 | 1203 | 132 | ||
| CN458042.1 | - | 4 - 5 | 382 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem87b
Genomic Map