Assay Details
| Gene Symbol: Tmem87b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028248.2 | NP_082524.2 | 8 - 9 | 878 | 122 | |
| GenBank mRNA | AK050203.1 | - | 8 - 9 | 870 | 122 | |
| BC031488.1 | - | 8 - 9 | 834 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem87b
Genomic Map