Assay Details
| Gene Symbol: Ckap2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181589.3 | NP_853620.1 | 3 - 4 | 231 | 70 | |
| GenBank mRNA | AK012040.1 | - | 3 - 4 | 183 | 70 | |
| AK153766.1 | - | 3 - 4 | 190 | 70 | ||
| BC053443.1 | - | 3 - 4 | 183 | 6333921 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ckap2l
Genomic Map