| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200039387.mm.1 |
NM_181589.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039387.mm.1 |
NM_181589.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039387.mm.1 |
TC0200004598.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200039390.mm.1 |
NM_181589.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039390.mm.1 |
NM_181589.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039390.mm.1 |
TC0200004598.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200039384.mm.1 |
NM_181589.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039384.mm.1 |
NM_181589.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039384.mm.1 |
TC0200004598.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200039386.mm.1 |
NM_181589.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039386.mm.1 |
NM_181589.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039386.mm.1 |
TC0200004598.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200039385.mm.1 |
NM_181589.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039385.mm.1 |
NM_181589.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039385.mm.1 |
TC0200004598.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200039388.mm.1 |
NM_181589.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039388.mm.1 |
NM_181589.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039388.mm.1 |
TC0200004598.mm.1 |
MTA TC |