Assay Details
| Gene Symbol: Cpsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016856.3 | NP_058552.1 | 12 - 13 | 2018 | 62 | |
| GenBank mRNA | AF012822.1 | - | 12 - 13 | 2004 | 62 | |
| BC007163.1 | - | 10 - 11 | 1676 | 62 | ||
| BC013628.1 | - | 12 - 13 | 1993 | 3497854 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf2
Genomic Map