Assay Details
| Gene Symbol: Cpsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016856.3 | NP_058552.1 | 4 - 5 | 606 | 158 | |
| GenBank mRNA | AF012822.1 | - | 4 - 5 | 592 | 158 | |
| AK054372.1 | - | 4 - 5 | 589 | 158 | ||
| AK140869.1 | - | 4 - 5 | 588 | 158 | ||
| AK141325.1 | - | 4 - 5 | 588 | 158 | ||
| AK147746.1 | - | 4 - 5 | 607 | 158 | ||
| BC007163.1 | - | 2 - 3 | 264 | 158 | ||
| BC013628.1 | - | 4 - 5 | 581 | 3497854 | 158 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf2
Genomic Map