Assay Details
| Gene Symbol: Fbln5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011812.4 | NP_035942.1 | 1 - 2 | 384 | 66 | |
| GenBank mRNA | AF112151.1 | - | 1 - 2 | 334 | 66 | |
| AK084895.1 | - | 1 - 2 | 225 | 66 | ||
| AK085170.1 | - | 1 - 2 | 214 | 66 | ||
| AK090129.1 | - | 1 - 2 | 323 | 66 | ||
| AK159189.1 | - | 1 - 2 | 147 | 66 | ||
| AK159471.1 | - | 1 - 2 | 367 | 66 | ||
| AK159960.1 | - | 1 - 2 | 354 | 66 | ||
| AK165018.1 | - | 1 - 2 | 177 | 66 | ||
| AK178699.1 | - | 1 - 2 | 64 | 66 | ||
| BC006636.1 | - | 1 - 2 | 74 | 3482574 | 66 | |
| BQ557120.1 | - | 1 - 2 | 384 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF112151.1, AK085170.1, AK090129.1, AK159471.1, AK159960.1, AK165018.1, BC006636.1, NM_011812.4 transcripts.
Set Membership: |
|

Fbln5
Genomic Map