Assay Details
| Gene Symbol: Fbln5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011812.4 | NP_035942.1 | 4 - 5 | 754 | 65 | |
| GenBank mRNA | AF112151.1 | - | 4 - 5 | 704 | 65 | |
| AK084895.1 | - | 4 - 5 | 595 | 65 | ||
| AK085170.1 | - | 4 - 5 | 584 | 65 | ||
| AK090129.1 | - | 4 - 5 | 693 | 65 | ||
| AK159189.1 | - | 4 - 5 | 517 | 65 | ||
| AK159471.1 | - | 4 - 5 | 737 | 65 | ||
| AK159960.1 | - | 4 - 5 | 724 | 65 | ||
| AK165018.1 | - | 4 - 5 | 547 | 65 | ||
| AK214662.1 | - | 1 - 2 | 46 | 65 | ||
| AK216833.1 | - | 1 - 2 | 46 | 65 | ||
| BC006636.1 | - | 4 - 5 | 444 | 3482574 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbln5
Genomic Map