Assay Details
| Gene Symbol: Fbln5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011812.4 | NP_035942.1 | 9 - 10 | 1362 | 85 | |
| GenBank mRNA | AF112151.1 | - | 9 - 10 | 1312 | 85 | |
| AK085170.1 | - | 9 - 10 | 1192 | 85 | ||
| AK090129.1 | - | 9 - 10 | 1301 | 85 | ||
| AK159189.1 | - | 9 - 10 | 1126 | 85 | ||
| AK159471.1 | - | 9 - 10 | 1345 | 85 | ||
| AK159960.1 | - | 9 - 10 | 1332 | 85 | ||
| AK165018.1 | - | 9 - 10 | 1155 | 85 | ||
| BC006636.1 | - | 9 - 10 | 1052 | 3482574 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbln5
Genomic Map