Assay Details
| Gene Symbol: Upf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081132.1 | NP_001074601.1 | 20 - 21 | 3783 | 88 | |
| GenBank mRNA | BC039278.1 | - | 9 - 10 | 1439 | 88 | |
| BC138358.1 | - | 20 - 21 | 3746 | 8861378 | 88 | |
| BC138359.1 | - | 20 - 21 | 3746 | 8861379 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Upf2
Genomic Map