Assay Details
| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021305.3 | NP_067280.1 | 3 - 4 | 227 | 85 | |
| GenBank mRNA | AF145254.1 | - | 3 - 4 | 163 | 85 | |
| AF222748.1 | - | 3 - 4 | 164 | 85 | ||
| AK017615.1 | - | 3 - 4 | 209 | 85 | ||
| AK077701.1 | - | 3 - 4 | 227 | 85 | ||
| AK136447.1 | - | 3 - 4 | 206 | 85 | ||
| AK160602.1 | - | 3 - 4 | 213 | 85 | ||
| AK161316.1 | - | 3 - 4 | 306 | 85 | ||
| BC005458.1 | - | 3 - 4 | 170 | 3494001 | 85 | |
More Information
Set Membership: |
|

Sec61a2
Genomic Map