Assay Details
| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021305.3 | NP_067280.1 | 5 - 6 | 438 | 71 | |
| GenBank mRNA | AF145254.1 | - | 5 - 6 | 374 | 71 | |
| AF222748.1 | - | 5 - 6 | 375 | 71 | ||
| AK017615.1 | - | 5 - 6 | 420 | 71 | ||
| AK077701.1 | - | 5 - 6 | 438 | 71 | ||
| AK136447.1 | - | 5 - 6 | 417 | 71 | ||
| AK160602.1 | - | 5 - 6 | 424 | 71 | ||
| BC005458.1 | - | 5 - 6 | 381 | 3494001 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec61a2
Genomic Map