Assay Details
| Gene Symbol: Camk1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290374.1 | NP_001277303.1 | 3 - 4 | 891 | 76 | |
| NM_001290375.1 | NP_001277304.1 | 4 - 5 | 966 | 76 | ||
| NM_001290376.1 | NP_001277305.1 | 4 - 5 | 482 | 76 | ||
| NM_177343.4 | NP_796317.2 | 4 - 5 | 966 | 76 | ||
| GenBank mRNA | AK052401.1 | - | 4 - 5 | 387 | 76 | |
| AK053173.1 | - | 4 - 5 | 720 | 76 | ||
| AK147616.1 | - | 4 - 5 | 967 | 76 | ||
| AK147657.1 | - | 4 - 5 | 677 | 76 | ||
| AK162754.1 | - | 4 - 5 | 681 | 76 | ||
| AK170123.1 | - | 3 - 4 | 650 | 76 | ||
| AK170777.1 | - | 4 - 5 | 731 | 76 | ||
| AY273822.1 | - | 4 - 5 | 482 | 76 | ||
| BC141413.1 | - | 4 - 5 | 466 | 9056026 | 76 | |
| BC141414.1 | - | 4 - 5 | 466 | 9056027 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camk1d
Genomic Map