Assay Details
| Gene Symbol: Timm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013899.2 | NP_038927.2 | 1 - 2 | 125 | 72 | |
| GenBank mRNA | AK008088.1 | - | 1 - 2 | 126 | 72 | |
| AK013546.1 | - | 1 - 2 | 55 | 72 | ||
| BC031448.1 | - | 1 - 2 | 3257567 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008088.1, BC031448.1, NM_013899.2 transcripts.
Set Membership: |
|

Timm10
Genomic Map