Assay Details
| Gene Symbol: Ssrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136081.2 | NP_001129553.1 | 7 - 8 | 1072 | 88 | |
| NM_182990.4 | NP_892035.2 | 7 - 8 | 1201 | 88 | ||
| GenBank mRNA | AK146446.1 | - | 7 - 8 | 1124 | 88 | |
| AK146585.1 | - | 7 - 8 | 1121 | 88 | ||
| AK146607.1 | - | 7 - 8 | 1104 | 88 | ||
| AK146691.1 | - | 7 - 8 | 1104 | 88 | ||
| AK147005.1 | - | 7 - 8 | 1132 | 88 | ||
| AK158552.1 | - | 7 - 8 | 1072 | 88 | ||
| BC004848.1 | - | 7 - 8 | 1068 | 88 | ||
| BC024835.1 | - | 4 - 5 | 571 | 88 | ||
| BC042502.1 | - | 7 - 8 | 1049 | 5005209 | 88 | |
| S50213.1 | - | 6 - 7 | 946 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ssrp1
Genomic Map