Assay Details
| Gene Symbol: Entpd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028093.1 | NP_082369.1 | 4 - 5 | 644 | 72 | |
| GenBank mRNA | AY364442.1 | - | 4 - 5 | 506 | 72 | |
| BC031143.1 | - | 4 - 5 | 640 | 4973216 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Entpd8
Genomic Map