Assay Details
| Gene Symbol: Pnpla7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146251.4 | NP_666363.3 | 9 - 10 | 1122 | 80 | |
| GenBank mRNA | AK038505.1 | - | 9 - 10 | 1008 | 80 | |
| AK088362.1 | - | 9 - 10 | 1068 | 80 | ||
| AK148380.1 | - | 9 - 10 | 1096 | 80 | ||
| AK170417.1 | - | 9 - 10 | 1068 | 80 | ||
| BC021312.1 | - | 9 - 10 | 983 | 80 | ||
| BC025621.1 | - | 5 - 6 | 535 | 80 | ||
| BC141365.1 | - | 9 - 10 | 906 | 9055978 | 80 | |
| BC141366.1 | - | 9 - 10 | 906 | 9055979 | 80 | |
| EU293208.1 | - | 8 - 9 | 793 | 80 | ||
| EU368675.1 | - | 8 - 9 | 793 | 80 | ||
| EU402948.2 | - | 8 - 9 | 793 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pnpla7
Genomic Map