Assay Details
| Gene Symbol: Pnpla7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146251.4 | NP_666363.3 | 19 - 20 | 2363 | 71 | |
| GenBank mRNA | AK038505.1 | - | 19 - 20 | 2249 | 71 | |
| AK088362.1 | - | 19 - 20 | 2309 | 71 | ||
| AK148380.1 | - | 19 - 20 | 2337 | 71 | ||
| AK170417.1 | - | 19 - 20 | 2309 | 71 | ||
| BC021312.1 | - | 18 - 19 | 2397 | 71 | ||
| BC025621.1 | - | 15 - 16 | 1776 | 71 | ||
| BC064003.1 | - | 2 - 3 | 127 | 71 | ||
| BC141365.1 | - | 19 - 20 | 2147 | 9055978 | 71 | |
| BC141366.1 | - | 19 - 20 | 2147 | 9055979 | 71 | |
| EU293208.1 | - | 18 - 19 | 2034 | 71 | ||
| EU368675.1 | - | 18 - 19 | 2034 | 71 | ||
| EU402948.2 | - | 18 - 19 | 2034 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pnpla7
Genomic Map