Assay Details
| Gene Symbol: Man1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029983.2 | NP_001025154.1 | 10 - 11 | 1559 | 72 | |
| GenBank mRNA | AK088523.1 | - | 5 - 6 | 845 | 72 | |
| BC006645.1 | - | 8 - 9 | 1175 | 72 | ||
| BC025500.1 | - | 2 - 3 | 211 | 72 | ||
| BC138550.1 | - | 10 - 11 | 1515 | 8861570 | 72 | |
| BC138551.1 | - | 10 - 11 | 1515 | 8861571 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Man1b1
Genomic Map