Assay Details
| Gene Symbol: Man1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029983.2 | NP_001025154.1 | 11 - 12 | 1757 | 61 | |
| GenBank mRNA | AK088523.1 | - | 6 - 7 | 1043 | 61 | |
| BC006645.1 | - | 9 - 10 | 1373 | 61 | ||
| BC025500.1 | - | 3 - 4 | 409 | 61 | ||
| BC138550.1 | - | 11 - 12 | 1713 | 8861570 | 61 | |
| BC138551.1 | - | 11 - 12 | 1713 | 8861571 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Man1b1
Genomic Map