Assay Details
| Gene Symbol: Man1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029983.2 | NP_001025154.1 | 2 - 3 | 346 | 95 | |
| GenBank mRNA | AK043541.1 | - | 2 - 3 | 327 | 95 | |
| AK207223.1 | - | 1 - 2 | 85 | 95 | ||
| BC138550.1 | - | 2 - 3 | 302 | 8861570 | 95 | |
| BC138551.1 | - | 2 - 3 | 302 | 8861571 | 95 | |
| BY732333.1 | - | 2 - 3 | 342 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Man1b1
Genomic Map