Assay Details
| Gene Symbol: Man1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029983.2 | NP_001025154.1 | 4 - 5 | 613 | 78 | |
| GenBank mRNA | AK043541.1 | - | 4 - 5 | 594 | 78 | |
| BC006645.1 | - | 2 - 3 | 229 | 78 | ||
| BC138550.1 | - | 4 - 5 | 569 | 8861570 | 78 | |
| BC138551.1 | - | 4 - 5 | 569 | 8861571 | 78 | |
| BY732333.1 | - | 4 - 5 | 609 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Man1b1
Genomic Map