Assay Details
| Gene Symbol: Man1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029983.2 | NP_001025154.1 | 5 - 6 | 724 | 79 | |
| GenBank mRNA | AK043541.1 | - | 5 - 6 | 705 | 79 | |
| AK178205.1 | - | 1 - 2 | 107 | 79 | ||
| BC006645.1 | - | 3 - 4 | 340 | 79 | ||
| BC138550.1 | - | 5 - 6 | 680 | 8861570 | 79 | |
| BC138551.1 | - | 5 - 6 | 680 | 8861571 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Man1b1
Genomic Map