Assay Details
| Gene Symbol: Man1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029983.2 | NP_001025154.1 | 8 - 9 | 1251 | 74 | |
| GenBank mRNA | AK088523.1 | - | 3 - 4 | 537 | 74 | |
| AK216461.1 | - | 1 - 2 | 42 | 74 | ||
| BC006645.1 | - | 6 - 7 | 867 | 74 | ||
| BC138550.1 | - | 8 - 9 | 1207 | 8861570 | 74 | |
| BC138551.1 | - | 8 - 9 | 1207 | 8861571 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Man1b1
Genomic Map