Assay Details
| Gene Symbol: Tmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290751.1 | NP_001277680.1 | 1 - 2 | 320 | 79 | |
| NM_025868.4 | NP_080144.1 | 2 - 3 | 506 | 79 | ||
| GenBank mRNA | AK009759.1 | - | 1 - 2 | 194 | 79 | |
| AK075750.1 | - | 2 - 3 | 508 | 79 | ||
| AK170587.1 | - | 1 - 2 | 203 | 79 | ||
| BC013544.1 | - | 1 - 2 | 188 | 4242418 | 79 | |
| BC110994.1 | - | 1 - 2 | 189 | 5349078 | 79 | |
| BQ840236.1 | - | 2 - 3 | 368 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tmx2
Genomic Map