Assay Details
| Gene Symbol: Prg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008920.4 | NP_032946.1 | 1 - 2 | 44 | 73 | |
| GenBank mRNA | AK010901.1 | - | 1 - 2 | 45 | 73 | |
| AK156715.1 | - | 1 - 2 | 46 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010901.1, AK156715.1, NM_008920.4 transcripts.
Set Membership: |
|

Prg2
Genomic Map