Assay Details
| Gene Symbol: Prg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016914.2 | NP_058610.1 | 2 - 3 | 225 | 90 | |
| GenBank mRNA | AF202533.1 | - | 2 - 3 | 225 | 90 | |
| AK158110.1 | - | 2 - 3 | 245 | 90 | ||
| BC104388.1 | - | 1 - 2 | 135 | 40050227 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prg3
Genomic Map