Assay Details
| Gene Symbol: Med19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025885.3 | NP_080161.1 | 2 - 3 | 514 | 89 | |
| GenBank mRNA | AK005399.1 | - | 2 - 3 | 482 | 89 | |
| AK010552.1 | - | 2 - 3 | 492 | 89 | ||
| AK160579.1 | - | 2 - 3 | 480 | 89 | ||
| AK168787.1 | - | 2 - 3 | 482 | 89 | ||
| BB611514.1 | - | 2 - 3 | 514 | 89 | ||
| BC025475.1 | - | 1 - 2 | 88 | 89 | ||
| BC044207.1 | - | 2 - 3 | 376 | 89 | ||
| BC044302.1 | - | 2 - 3 | 376 | 89 | ||
| BC055404.1 | - | 2 - 3 | 469 | 89 | ||
| BC082559.1 | - | 2 - 3 | 479 | 30606373 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med19
Genomic Map