Assay Details
| Gene Symbol: Ssna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023464.2 | NP_075953.1 | 2 - 3 | 322 | 89 | |
| GenBank mRNA | AB041656.1 | - | 2 - 3 | 316 | 89 | |
| AK088418.1 | - | 2 - 3 | 254 | 89 | ||
| AK158412.1 | - | 439 | 89 | |||
| BC023033.1 | - | 2 - 3 | 285 | 89 | ||
| BC037081.1 | - | 2 - 3 | 299 | 5098653 | 89 | |
| BC048360.1 | - | 2 - 3 | 300 | 5067090 | 89 | |
| BY703613.1 | - | 2 - 3 | 323 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21071030 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21071032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21071033 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21071031 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321965 | AK158412.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321965 | AK208309.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321965 | BC023033.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321965 | BC037081.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321965 | BC048360.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321965 | AK158412.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321965 | AK208309.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321965 | BC023033.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321965 | BC037081.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321965 | BC048360.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285051 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145584 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321965 | AK158412.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321965 | AK208309.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321965 | BC023033.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321965 | BC037081.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321965 | BC048360.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321965 | AK158412.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321965 | AK208309.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321965 | BC023033.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321965 | BC037081.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321965 | BC048360.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21071022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21071030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21071034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21071027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21071029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21071023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21071033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21071031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21071028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21071026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21071019 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145584 | - | - |
| Whole Mouse Genome G4122A | A_51_P321965 | AK158412.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321965 | AK208309.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321965 | BC023033.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321965 | BC037081.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321965 | BC048360.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321965 | AK158412.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321965 | AK208309.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321965 | BC023033.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321965 | BC037081.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321965 | BC048360.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140274 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145584 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01825815 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01825825 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01825830 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01825814 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21071030 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21071032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21071031 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21071033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21071022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01825830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01825820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01825817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21071031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01825831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01825827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01825814 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10480673 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480675 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480674 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448643_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10480672 | - | - |
| Mouse Genome 430 2.0 Array | 1448643_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448643_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68475_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68475_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200025869.mm.1 | NM_023464.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025869.mm.1 | NM_023464.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025869.mm.1 | TC0200003141.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200025868.mm.1 | NM_023464.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025868.mm.1 | NM_023464.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025868.mm.1 | TC0200003141.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448643_PM_at | - | - |
| Mouse Expression Set 430 | 1448643_at | - | - |
| Murine Genome U74v2 Set | 93805_PM_at | - | - |
| Murine Genome U74v2 Set | 93805_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448643_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448643_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448643_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448643_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161639 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161639 | - | - |
| mouse-6 v1.1 (Illumina) | GI_12963686-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161639 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161639 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_12963686-S | - | - |

Ssna1
Genomic Map