Assay Details
| Gene Symbol: Map2k4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009157.4 | NP_033183.1 | 5 - 6 | 681 | 159 | |
| GenBank mRNA | AK036928.1 | - | 5 - 6 | 685 | 159 | |
| AK044534.1 | - | 5 - 6 | 673 | 159 | ||
| AK044698.1 | - | 5 - 6 | 673 | 159 | ||
| AK139777.1 | - | 6 - 7 | 704 | 159 | ||
| BC029833.1 | - | 3 - 4 | 432 | 3962163 | 159 | |
| U18310.1 | - | 5 - 6 | 656 | 159 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k4
Genomic Map