Assay Details
| Gene Symbol: Map2k4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009157.4 | NP_033183.1 | 7 - 8 | 866 | 58 | |
| GenBank mRNA | AK036928.1 | - | 7 - 8 | 870 | 58 | |
| AK044534.1 | - | 7 - 8 | 858 | 58 | ||
| AK044698.1 | - | 7 - 8 | 858 | 58 | ||
| AK139777.1 | - | 8 - 9 | 889 | 58 | ||
| BC029833.1 | - | 5 - 6 | 617 | 3962163 | 58 | |
| U18310.1 | - | 7 - 8 | 841 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k4
Genomic Map