Assay Details
| Gene Symbol: Grb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016719.1 | NP_057928.1 | 12 - 13 | 1500 | 99 | |
| GenBank mRNA | AF155647.1 | - | 12 - 13 | 1500 | 99 | |
| AK147041.1 | - | 12 - 13 | 1509 | 99 | ||
| BC021820.1 | - | 5 - 6 | 386 | 99 | ||
| BC138581.1 | - | 12 - 13 | 1406 | 8861601 | 99 | |
| BC145913.1 | - | 12 - 13 | 1406 | 40131216 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grb14
Genomic Map