Assay Details
| Gene Symbol: Katnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024254.3 | NP_077216.1 | 4 - 5 | 586 | 85 | |
| GenBank mRNA | AK010654.1 | - | 4 - 5 | 565 | 85 | |
| AK036861.1 | - | 4 - 5 | 586 | 85 | ||
| AK151464.1 | - | 4 - 5 | 572 | 85 | ||
| AK152570.1 | - | 4 - 5 | 551 | 85 | ||
| AK168639.1 | - | 4 - 5 | 584 | 85 | ||
| BC003216.1 | - | 4 - 5 | 542 | 3157982 | 85 | |
| BC012402.1 | - | 4 - 5 | 542 | 3157982 | 85 | |
| BC080661.1 | - | 4 - 5 | 542 | 4460520 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Katnbl1
Genomic Map