Assay Details
| Gene Symbol: Snx9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025664.5 | NP_079940.2 | 11 - 12 | 1344 | 74 | |
| GenBank mRNA | AK012520.1 | - | 6 - 7 | 636 | 74 | |
| AK139802.1 | - | 11 - 12 | 1378 | 74 | ||
| AK149739.1 | - | 11 - 12 | 1358 | 74 | ||
| AK149849.1 | - | 11 - 12 | 1398 | 74 | ||
| AK155830.1 | - | 1379 | 74 | |||
| AK157517.1 | - | 11 - 12 | 1360 | 74 | ||
| AK170339.1 | - | 1360 | 74 | |||
| BC014814.1 | - | 11 - 12 | 1344 | 3257422 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snx9
Genomic Map