Assay Details
| Gene Symbol: Serac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111017.1 | NP_001104487.1 | 13 - 14 | 1528 | 70 | |
| NM_177311.4 | NP_796285.2 | 12 - 13 | 1438 | 70 | ||
| GenBank mRNA | AK019677.1 | - | 12 - 13 | 1427 | 70 | |
| AK154605.1 | - | 13 - 14 | 1479 | 70 | ||
| AK155571.1 | - | 13 - 14 | 1531 | 70 | ||
| AK170498.1 | - | 13 - 14 | 1479 | 70 | ||
| BC119594.1 | - | 13 - 14 | 1517 | 40059066 | 70 | |
| BC119595.1 | - | 13 - 14 | 1517 | 40059071 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Serac1
Genomic Map