Assay Details
| Gene Symbol: Prpf4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013830.2 | NP_038858.2 | 1 - 2 | 198 | 126 | |
| GenBank mRNA | AF283466.1 | - | 1 - 2 | 196 | 126 | |
| AK020579.1 | - | 1 - 2 | 68 | 126 | ||
| AK021274.1 | - | 1 - 2 | 82 | 126 | ||
| AK078611.1 | - | 1 - 2 | 164 | 126 | ||
| AK083926.1 | - | 1 - 2 | 82 | 126 | ||
| AK165391.1 | - | 1 - 2 | 189 | 126 | ||
| BC042521.1 | - | 1 - 2 | 113 | 126 | ||
| BC059713.1 | - | 1 - 2 | 170 | 126 | ||
| BC080864.1 | - | 1 - 2 | 174 | 126 | ||
| BC141272.1 | - | 1 - 2 | 124 | 9055884 | 126 | |
| BC141273.1 | - | 1 - 2 | 124 | 9055885 | 126 | |
| CJ046150.1 | - | 1 - 2 | 199 | 126 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF283466.1, AK020579.1, AK021274.1, AK078611.1, AK083926.1, BC059713.1, BC141272.1, BC141273.1, NM_013830.2 transcripts.
Set Membership: |
|

Prpf4b
Genomic Map