| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200029287.mm.1 |
NM_146122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029287.mm.1 |
NM_146122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029287.mm.1 |
TC0200003407.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200029289.mm.1 |
NM_146122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029289.mm.1 |
NM_146122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029289.mm.1 |
TC0200003407.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200029295.mm.1 |
NM_146122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029295.mm.1 |
NM_146122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029295.mm.1 |
TC0200003407.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200029297.mm.1 |
NM_146122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029297.mm.1 |
NM_146122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029297.mm.1 |
TC0200003407.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200029300.mm.1 |
NM_146122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029300.mm.1 |
NM_146122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029300.mm.1 |
TC0200003407.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200029298.mm.1 |
NM_146122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029298.mm.1 |
NM_146122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200029298.mm.1 |
TC0200003407.mm.1 |
MTA TC |