Assay Details
| Gene Symbol: Hc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010406.2 | NP_034536.1 | 19 - 20 | 2486 | 80 | |
| GenBank mRNA | M35525.1 | - | 19 - 20 | 2441 | 80 | |
| M35526.1 | - | 19 - 20 | 2439 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hc
Genomic Map