Assay Details
| Gene Symbol: Hc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010406.2 | NP_034536.1 | 3 - 4 | 475 | 155 | |
| GenBank mRNA | AK080166.1 | - | 3 - 4 | 474 | 155 | |
| M35525.1 | - | 3 - 4 | 430 | 155 | ||
| M35526.1 | - | 3 - 4 | 430 | 155 | ||
More Information
Set Membership: |
|

Hc
Genomic Map