Assay Details
| Gene Symbol: 4930568D16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029463.1 | NP_083739.1 | 1 - 2 | 102 | 127 | |
| GenBank mRNA | AK016248.1 | - | 1 - 2 | 103 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016248.1, NM_029463.1 transcripts.
Set Membership: |
|

4930568D16Rik
Genomic Map