Assay Details
| Gene Symbol: Amn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017204.1 | - | 1 - 2 | 69 | 113 | |
| BC087954.1 | - | 1 - 2 | 68 | 30119116 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC087954.1 transcripts.
Set Membership: |
|

Amn
Genomic Map