Assay Details
| Gene Symbol: Cutal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021330.1 | - | 4 - 5 | 368 | 118 | |
| AK033810.1 | - | 4 - 5 | 446 | 118 | ||
| AK085616.1 | - | 4 - 5 | 407 | 118 | ||
More Information
Set Membership: |
|

Cutal
Genomic Map